Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2'-methyldiphenyl Ether 97.0+%, TCI America™
SDP

Catalog No. A100825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A100825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A100825G Supplier TCI America Supplier No. A100825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3840-18-4
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00025167
InChI Key JYJPXACGURQSCB-UHFFFAOYSA-N
Synonym 2-Aminophenyl o-Tolyl Ether, 2-(o-Tolyloxy)aniline
PubChem CID 19692
IUPAC Name 2-(2-methylphenoxy)aniline
SMILES CC1=CC=CC=C1OC2=CC=CC=C2N

Specifications

Boiling Point 196°C
Quantity 25 g
Formula Weight 199.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amino-2′-methyldiphenyl Ether
RTECSNumber BY9750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.