accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38404-42-1

IUPAC Name: methyl 3,4-dimethylbenzoate
Synonyms: methyl 3,4-dimethylbenzoate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: PTSSKYUSCIALKU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)