Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,4-dimethylbenzoate, 98%, Thermo Scientific™

Catalog No. AAB2002418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2002418 50 g
AAB2002409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2002418 Supplier Thermo Scientific Chemicals Supplier No. B2002418

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used to produce 3,4-Dimethyl-benzyl alcohol.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38404-42-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00219861
InChI Key PTSSKYUSCIALKU-UHFFFAOYSA-N
Synonym 3,4-dimethylbenzoic acid methyl ester, benzoic acid, 3,4-dimethyl-, methyl ester, acmc-20akhv, 4-methoxycarbonyl-o-xylene, #, methyl ester of 3,4-dimethylbenzoic acid
PubChem CID 520902
IUPAC Name methyl 3,4-dimethylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)C

Specifications

Melting Point 35°C to 37°C
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 164.21
Percent Purity 98%
Chemical Name or Material Methyl 3,4-dimethylbenzoate
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.