CAS RN 38438-11-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dibutyltin Bis(trifluoromethanesulfonate) 98.0+%, TCI America™
CAS: 38438-11-8 Molecular Formula: C10H18F6O6S2Sn MDL Number: MFCD00671529 Synonym: Dibutyltin Ditriflate