Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyltin Bis(trifluoromethanesulfonate) 98.0+%, TCI America™
SDP

Catalog No. D24665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24665G Supplier TCI America Supplier No. D24665G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38438-11-8
Molecular Formula C10H18F6O6S2Sn
MDL Number MFCD00671529
Synonym Dibutyltin Ditriflate

Specifications

Color Red-White
Quantity 5 g
UN Number 2923
Formula Weight 531.07
Percent Purity ≥98.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Dibutyltin Bis(trifluoromethanesulfonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.