accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3845-76-9

IUPAC Name: N-[3-(dimethylamino)propyl]prop-2-enamide
Synonyms: N-[3-(dimethylamino)propyl]prop-2-enamide
Molecular Weight (g/mol): 156.23
Molecular Formula: C8H16N2O
InChi Key: ADTJPOBHAXXXFS-UHFFFAOYSA-N
SMILES: CN(C)CCCNC(=O)C=C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results