Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[3-(Dimethylamino)propyl]acrylamide (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. D1785500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1785500G 500 g
D178525G 25 g
D1785100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1785500G Supplier TCI America Supplier No. D1785500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3845-76-9
Molecular Formula C8H16N2O
Molecular Weight (g/mol) 156.23
MDL Number MFCD00059902
InChI Key ADTJPOBHAXXXFS-UHFFFAOYSA-N
PubChem CID 77452
IUPAC Name N-[3-(dimethylamino)propyl]prop-2-enamide
SMILES CN(C)CCCNC(=O)C=C

Specifications

Color Yellow
Boiling Point 142°C
Quantity 500 g
Formula Weight 156.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-[3-(Dimethylamino)propyl]acrylamide (stabilized with MEHQ)
EINECSNumber (2)-1013
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.