accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOOH3C

CAS RN 38464-04-9

IUPAC Name: 2-(3,4-diethoxyphenyl)acetic acid
Synonyms: (3,4-diethoxyphenyl)acetic acid
Molecular Weight (g/mol): 224.26
Molecular Formula: C12H16O4
InChi Key: FIKUHWAANCXBGJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(CC(O)=O)C=C1OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results