Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Diethoxyphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D34195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34195G Supplier TCI America Supplier No. D34195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38464-04-9
Molecular Formula C12H16O4
Molecular Weight (g/mol) 224.26
MDL Number MFCD00040785
InChI Key FIKUHWAANCXBGJ-UHFFFAOYSA-N
PubChem CID 579813
IUPAC Name 2-(3,4-diethoxyphenyl)acetic acid
SMILES CCOC1=CC=C(CC(O)=O)C=C1OCC

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 5 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Diethoxyphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.