accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOF

CAS RN 385380-74-5

IUPAC Name: 1-(2-fluoro-4-nitrophenyl)pyrrolidine
Synonyms: 1-(2-fluoro-4-nitrophenyl)pyrrolidine
Molecular Weight (g/mol): 210.21
Molecular Formula: C10H11FN2O2
InChi Key: SMKXLBISYVJESG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results