CAS RN 38557-72-1
CAS RN 38557-72-1
IUPAC Name:
2-chloro-3,5-dimethylpyrazine
Synonyms:
2-chloro-3,5-dimethylpyrazine
Molecular Weight (g/mol):
142.59
Molecular Formula:
C6H7ClN2
InChi Key:
BTGGHNHGPURMEO-UHFFFAOYSA-N
SMILES:
CC1=CN=C(Cl)C(C)=N1
1
–
1
of
1
results
1
2-Chloro-3,5-dimethylpyrazine 95.0+%, TCI America™

CAS: 38557-72-1 Molecular Formula: C6H7ClN2 Molecular Weight (g/mol): 142.59 MDL Number: MFCD00126945 InChI Key: BTGGHNHGPURMEO-UHFFFAOYSA-N Synonym: 2-chloro 3,5-dimethyl pyarazine,2-chloro-3,5-dimethyl-1,4-diazine,2-chloro-3,5-dimethyl-pyrazine,3,5-dimethyl-2-chloropyrazine,2-chloro 3,5-dimethyl pyrazine,2-chloro-3,5-dimethyl pyrazine,pyrazine,2-chloro-3,5-dimethyl,2-chloranyl-3,5-dimethyl-pyrazine,pyrazine, 2-chloro-3,5-dimethyl,3-chloro-2,6-dimethylpyrazine PubChem CID: 11672680 IUPAC Name: 2-chloro-3,5-dimethylpyrazine SMILES: CC1=CN=C(Cl)C(C)=N1