Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3,5-dimethylpyrazine 95.0+%, TCI America™
SDP

Catalog No. C31181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31181G Supplier TCI America Supplier No. C31181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38557-72-1
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00126945
InChI Key BTGGHNHGPURMEO-UHFFFAOYSA-N
Synonym 2-chloro 3,5-dimethyl pyarazine, 2-chloro-3,5-dimethyl-1,4-diazine, 2-chloro-3,5-dimethyl-pyrazine, 3,5-dimethyl-2-chloropyrazine, 2-chloro 3,5-dimethyl pyrazine, 2-chloro-3,5-dimethyl pyrazine, pyrazine,2-chloro-3,5-dimethyl, 2-chloranyl-3,5-dimethyl-pyrazine, pyrazine, 2-chloro-3,5-dimethyl, 3-chloro-2,6-dimethylpyrazine
PubChem CID 11672680
IUPAC Name 2-chloro-3,5-dimethylpyrazine
SMILES CC1=CN=C(Cl)C(C)=N1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 1 g
Formula Weight 142.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3,5-dimethylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.