accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38565-53-6

IUPAC Name: 2-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononyl)oxirane
Synonyms: 2-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononyl)oxirane
Molecular Weight (g/mol): 476.13
Molecular Formula: C11H5F17O
InChi Key: HMXSIEIEXLGIET-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results