Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxy-1H,1H,2H,3H,3H-heptadecafluoroundecane 98.0+%, TCI America™
SDP

Catalog No. E046210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E046210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E046210G Supplier TCI America Supplier No. E046210G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38565-53-6
Molecular Formula C11H5F17O
Molecular Weight (g/mol) 476.13
MDL Number MFCD00042362
InChI Key HMXSIEIEXLGIET-UHFFFAOYNA-N
Synonym 3-Perfluorooctyl-1,2-epoxypropane, 3-(Perfluoro-n-octyl)propenoxide
PubChem CID 571982
IUPAC Name 2-(2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-heptadecafluorononyl)oxirane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1

Specifications

Color Colorless
Boiling Point 87°C
Quantity 10 g
Formula Weight 476.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxy-1H,1H,2H,3H,3H-heptadecafluoroundecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.