accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OH3CCH3NO

CAS RN 38568-24-0

IUPAC Name: [2-(4-methoxy-4-oxobutyl)-4,4-dimethyl-2-tridecyl-1,3-oxazolidin-3-yl]oxidanyl
Synonyms: 2-(4-methoxy-4-oxobutyl)-4,4-dimethyl-2-tridecyl-1,3-oxazolidin-3-yloxidanyl
Molecular Weight (g/mol): 398.61
Molecular Formula: C23H44NO4
InChi Key: XKWHHWBOJRTEHX-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCC1(CCCC(=O)OC)OCC(C)(C)N1[O]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)