Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle5-DOXYL-stearate, free radical, Thermo Scientific™

Catalog No. AC225130100
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225130100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225130100 Supplier Thermo Scientific Chemicals Supplier No. 225130100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Polymerization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38568-24-0
Molecular Formula C23H44NO4
Molecular Weight (g/mol) 398.608
InChI Key XKWHHWBOJRTEHX-UHFFFAOYSA-N
Synonym methyl 5-doxyl-stearate, methyl 5-doxyl-stearate, free radical, 5-doxylstearic acid methyl ester, 5-doxylstearate methyl ester, methyl 5-doxylstearate, methyl 4-3, 5-mesl, 2-4-methoxy-4-oxobutyl-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxy, 3-oxazolidinyloxy, 2-4-methoxy-4-oxobutyl-4,4-dimethyl-2-tridecyl, 2-4-methoxy-4-oxobutyl-4,4-dimethyl-2-tridecyl-3-oxazolidinyloxy, free radical
PubChem CID 170076
IUPAC Name methyl 4-(3-$l^{1}-oxidanyl-4,4-dimethyl-2-tridecyl-1,3-oxazolidin-2-yl)butanoate
SMILES CCCCCCCCCCCCCC1(N(C(CO1)(C)C)[O])CCCC(=O)OC

Specifications

Quantity 10 mg
Formula Weight 398.59
Chemical Name or Material Methyle5-DOXYL-stearate, free radical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.