accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3858-89-7

HClH3CNCH3OONH2Cl

CAS RN 3858-89-7

IUPAC Name: 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride
Synonyms: chloroprocaine hcl
Molecular Weight (g/mol): 307.22
Molecular Formula: C13H20Cl2N2O2
InChi Key: SZKQYDBPUCZLRX-UHFFFAOYSA-N
SMILES: Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results