Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chloroprocaine hydrochloride

Catalog No. AC465372500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465372500 250 mg
AC465370010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465372500 Supplier Thermo Scientific Chemicals Supplier No. 465372500
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3858-89-7
Molecular Formula C13H20Cl2N2O2
Molecular Weight (g/mol) 307.22
InChI Key SZKQYDBPUCZLRX-UHFFFAOYSA-N
IUPAC Name 2-(diethylamino)ethyl 4-amino-2-chlorobenzoate hydrochloride
SMILES Cl.CCN(CC)CCOC(=O)C1=CC=C(N)C=C1Cl

Specifications

Melting Point 173.0°C to 178.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 307.22
Chemical Name or Material Chloroprocaine hydrochloride
EINECSNumber 223-371-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.