accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CCH3ClCH3CH3

CAS RN 38580-86-8

IUPAC Name: 2-(chloromethyl)-1,3,5-tris(propan-2-yl)benzene
Synonyms: 2-(chloromethyl)-1,3,5-triisopropylbenzene
Molecular Weight (g/mol): 252.83
Molecular Formula: C16H25Cl
InChi Key: CQHQLMOCMQMZPP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(CCl)C(=C1)C(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results