Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triisopropylbenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. T19895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19895G Supplier TCI America Supplier No. T19895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38580-86-8
Molecular Formula C16H25Cl
Molecular Weight (g/mol) 252.826
MDL Number MFCD03701106
InChI Key CQHQLMOCMQMZPP-UHFFFAOYSA-N
Synonym 2-Chloromethyl-1,3,5-triisopropylbenzene
PubChem CID 12480672
IUPAC Name 2-(chloromethyl)-1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)CCl)C(C)C

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 252.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triisopropylbenzyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.