accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 385810-21-9

IUPAC Name: 3-(4-butylphenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one
Synonyms: 3-(4-butylphenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 280.37
Molecular Formula: C19H20O2
InChi Key: LFLVYHVGNKJGBV-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=CC(=O)C2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results