Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butyl-4'-hydroxychalcone 98.0+%, TCI America™
SDP

Catalog No. B27411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B27411G Supplier TCI America Supplier No. B27411G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 385810-21-9
Molecular Formula C19H20O2
Molecular Weight (g/mol) 280.367
MDL Number MFCD08276309
InChI Key LFLVYHVGNKJGBV-UHFFFAOYSA-N
Synonym 2-(4-Butylbenzylidene)-4′C-hydroxyacetophenone, 4-(4-Butylcinnamoyl)phenol
PubChem CID 44629782
IUPAC Name 3-(4-butylphenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one
SMILES CCCCC1=CC=C(C=C1)C=CC(=O)C2=CC=C(C=C2)O

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 1 g
Formula Weight 280.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butyl-4′-hydroxychalcone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.