accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2FFF

CAS RN 3862-73-5

IUPAC Name: 2,3,4-trifluoroaniline
Synonyms: benzenamine, 2,3,4-trifluoro-
Molecular Weight (g/mol): 147.1
Molecular Formula: C6H4F3N
InChi Key: WRDGNXCXTDDYBZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C(F)=C1F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)