Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trifluoroaniline 98.0+%, TCI America™
SDP

Catalog No. T13855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13855G 5 g
T138525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13855G Supplier TCI America Supplier No. T13855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3862-73-5
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00011737
InChI Key WRDGNXCXTDDYBZ-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzenamine, benzenamine, 2,3,4-trifluoro, 2,3,4-trifluoraniline, 2,3,4-trifluoro-phenylamine, aniline, 2,3,4-trifluoro, 2,3,4-trifluoro aniline, 2,3,4-trifluorophenylamine, pubchem2297, acmc-1clgf, 2.3.4-trifluoroaniline
PubChem CID 77468
IUPAC Name 2,3,4-trifluoroaniline
SMILES C1=CC(=C(C(=C1N)F)F)F

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
UN Number 3082
Formula Weight 147.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4-Trifluoroaniline
RTECSNumber CY1211500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.