accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONCS

CAS RN 38663-85-3

IUPAC Name: 1-isothiocyanato-2-methoxyethane
Synonyms: 1-isothiocyanato-2-methoxyethane
Molecular Weight (g/mol): 117.17
Molecular Formula: C4H7NOS
InChi Key: HTSAVXAFEVUJQE-UHFFFAOYSA-N
SMILES: COCCN=C=S
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)