Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethyl isothiocyanate, 98%, Thermo Scientific™

Catalog No. AC333010050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC333010050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC333010050 Supplier Thermo Scientific Chemicals Supplier No. 333010050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38663-85-3
Molecular Formula C4H7NOS
Molecular Weight (g/mol) 117.166
InChI Key HTSAVXAFEVUJQE-UHFFFAOYSA-N
Synonym 2-methoxyethyl isothiocyanate, ethane, 1-isothiocyanato-2-methoxy, 2-methoxyethylisothiocyanate, 1-isothiocyanato-2-methoxy-ethane, 1-isothiocyanato-2-methoxyethane, 2-methoxyethanisothiocyanate, acmc-1aezw, 2-methoxy ethyl isothiocyanate, #, ethane,1-isothiocyanato-2-methoxy
PubChem CID 123475
IUPAC Name 1-isothiocyanato-2-methoxyethane
SMILES COCCN=C=S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.5110 to 1.5130 (20°C, 589nm)
Quantity 5 g
Formula Weight 117.2
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Methoxyethyl isothiocyanate
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.