accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38668-46-1

IUPAC Name: 6-[2-(2-methyl-1H-imidazol-1-yl)ethyl]-1,3,5-triazine-2,4-diamine
Synonyms: 6-[2-(2-methylimidazol-1-yl)ethyl]-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol): 219.25
Molecular Formula: C9H13N7
InChi Key: DRQWQDPSMJHCCM-UHFFFAOYSA-N
SMILES: CC1=NC=CN1CCC1=NC(N)=NC(N)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results