Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-[2-(2-methyl-1-imidazolyl)ethyl]-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D323825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D323825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D323825G Supplier TCI America Supplier No. D323825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38668-46-1
Molecular Formula C9H13N7
Molecular Weight (g/mol) 219.252
InChI Key DRQWQDPSMJHCCM-UHFFFAOYSA-N
PubChem CID 162320
IUPAC Name 6-[2-(2-methylimidazol-1-yl)ethyl]-1,3,5-triazine-2,4-diamine
SMILES CC1=NC=CN1CCC2=NC(=NC(=N2)N)N

Specifications

Melting Point 253°C
Color White-Yellow
Quantity 25 g
Formula Weight 219.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-[2-(2-methyl-1-imidazolyl)ethyl]-1,3,5-triazine
EINECSNumber (5)-1009
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.