accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOPOONOOONOO

CAS RN 3871-20-3

IUPAC Name: tris(4-nitrophenyl) phosphate
Synonyms: tris(4-nitrophenyl) phosphate
Molecular Weight (g/mol): 461.28
Molecular Formula: C18H12N3O10P
InChi Key: RZSPPBDBWOJRII-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OP(=O)(OC2=CC=C(C=C2)[N+]([O-])=O)OC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results