Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-nitrophenyl) Phosphate 98.0+%, TCI America™
SDP

Catalog No. T100225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T100225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T100225G Supplier TCI America Supplier No. T100225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3871-20-3
Molecular Formula C18H12N3O10P
Molecular Weight (g/mol) 461.279
MDL Number MFCD00024649
InChI Key RZSPPBDBWOJRII-UHFFFAOYSA-N
Synonym Phosphoric Acid Tris(4-nitrophenyl) Ester
PubChem CID 77473
IUPAC Name tris(4-nitrophenyl) phosphate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(OC2=CC=C(C=C2)[N+](=O)[O-])OC3=CC=C(C=C3)[N+](=O)[O-]

Specifications

Melting Point 155°C
Quantity 25 g
Formula Weight 461.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-nitrophenyl) Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.