accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CCH3H3CCH3CH3H3CHLi

CAS RN 38721-52-7

IUPAC Name: lithium(1+) tris(butan-2-yl)boranuide
Synonyms: L-selectride
Molecular Weight (g/mol): 190.11
Molecular Formula: C12H28BLi
InChi Key: ACJKNTZKEFMEAK-UHFFFAOYNA-N
SMILES: [Li+].CCC(C)[BH-](C(C)CC)C(C)CC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)