Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-Selectride™ Solution

Catalog No. 1784978L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
8 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-849-78L 8 L
17-849-7800ML 800 mL
17-849-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 17-849-78L Supplier Sigma Aldrich Supplier No. 1784978L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

L-Selectride™ is an air and water sensitive solution mainly used as a bulky stereoselective hydride reducing agent.L-Selectride(TM) solution may be used in the O-demethylation and N-demethylation of opium alkaloids and their derivatives.Selectride is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 38721-52-7
Density 0.89 g/mL at 25°C
Molecular Formula C12H28BLi
Linear Formula Li[CH(CH3)CH2CH3]3BH
Quantity 8 L
MDL Number MFCD00011708
Synonym Lithium tri-sec-butylborohydride solution
Molecular Weight (g/mol) 190.1 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.