accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 3875-51-2

IUPAC Name: (propan-2-yl)cyclopentane
Synonyms: isopropylcyclopentane
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: TVSBRLGQVHJIKT-UHFFFAOYSA-N
SMILES: CC(C)C1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results