Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylcyclopentane 98.0+%, TCI America™
SDP

Catalog No. I06471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06471G Supplier TCI America Supplier No. I06471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3875-51-2
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00045422
InChI Key TVSBRLGQVHJIKT-UHFFFAOYSA-N
PubChem CID 19751
IUPAC Name (propan-2-yl)cyclopentane
SMILES CC(C)C1CCCC1

Specifications

Color Colorless
Boiling Point 126°C
Quantity 1 g
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropylcyclopentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.