accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOO

CAS RN 38768-08-0

IUPAC Name: 7-oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzene-1-sulfonate
Synonyms: 7-oxocyclohepta-1,3,5-trien-1-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 276.31
Molecular Formula: C14H12O4S
InChi Key: AWUMFQREGHLKJB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=CC=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results