accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3877-15-4

IUPAC Name: 1-(methylsulfanyl)propane
Synonyms: N-propylmethylsulfide
Molecular Weight (g/mol): 90.18
Molecular Formula: C4H10S
InChi Key: ZOASGOXWEHUTKZ-UHFFFAOYSA-N
SMILES: CCCSC
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)