Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Propyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. M228525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M228525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M228525G Supplier TCI America Supplier No. M228525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3877-15-4
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00015219
InChI Key ZOASGOXWEHUTKZ-UHFFFAOYSA-N
Synonym methyl propyl sulfide, propane, 1-methylthio, sulfide, methyl propyl, methyl propyl sulphide, unii-nxa8rq27b3, 1-methylthio propane, 1-methylthio-propane, n-propylmethylsulfide, nxa8rq27b3, methyl n-propyl sulfide
PubChem CID 19754
IUPAC Name 1-(methylsulfanyl)propane
SMILES CCCSC

Specifications

Melting Point -113°C
Color Yellow
Boiling Point 97°C
Quantity 25 g
UN Number 1993
Formula Weight 90.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Propyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.