accessibility menu, dialog, popup

UserName

Viewing profile as user:
SeP

CAS RN 3878-44-2

IUPAC Name: triphenyl-λ⁵-phosphaneselone
Synonyms: triphenylphosphine selenide
Molecular Weight (g/mol): 341.26
Molecular Formula: C18H15PSe
InChi Key: ZFVJLNKVUKIPPI-UHFFFAOYSA-N
SMILES: [Se]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results