Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylphosphine Selenide 98.0+%, TCI America™
SDP

Catalog No. T18195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18195G Supplier TCI America Supplier No. T18195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3878-44-2
Molecular Formula C18H15PSe
Molecular Weight (g/mol) 341.26
MDL Number MFCD00013477
InChI Key ZFVJLNKVUKIPPI-UHFFFAOYSA-N
Synonym triphenylphosphine selenide, phosphine selenide, triphenyl, triphenylphosphine selenide 99+%, triphenyl-??-phosphaneselone, triphenyl selanylidene
PubChem CID 298720
IUPAC Name triphenyl-λ⁵-phosphaneselone
SMILES [Se]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 189°C
Color White
Quantity 5 g
UN Number 3283
Formula Weight 341.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylphosphine Selenide
RTECSNumber SZ2455000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.