accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3879-08-1

IUPAC Name: 4-hydroxybutanehydrazide
Synonyms: 4-hydroxybutanehydrazide
Molecular Weight (g/mol): 118.14
Molecular Formula: C4H10N2O2
InChi Key: WOXQFPPKZBOSTN-UHFFFAOYSA-N
SMILES: NNC(=O)CCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results