Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybutyric acid hydrazide, 98%, Thermo Scientific™

Catalog No. AAA1235122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1235122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1235122 Supplier Thermo Scientific Chemicals Supplier No. A1235122

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3879-08-1
Molecular Formula C4H10N2O2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00051702
InChI Key WOXQFPPKZBOSTN-UHFFFAOYSA-N
IUPAC Name 4-hydroxybutanehydrazide

Specifications

Melting Point 91°C to 95°C
Quantity 100 g
Beilstein 1749234
Synonym 4-hydroxybutyric acid hydrazide, 4-hydroxybutanohydrazide, butanoic acid, 4-hydroxy-, hydrazide, 4-???butan-1-ol, acmc-1cts6, 4-hydroxybutanoyl hydrazine, 4-hydroxybutanoic acid hydrazide, butanoic acid,4-hydroxy-, hydrazide
SMILES C(CC(=O)NN)CO
PubChem CID 220659
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 4-Hydroxybutyric acid hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.