accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2

CAS RN 38793-01-0

IUPAC Name: 4-pentylcyclohexan-1-amine
Synonyms: 4-pentylcyclohexan-1-amine
Molecular Weight (g/mol): 169.31
Molecular Formula: C11H23N
InChi Key: NQEVYZKSODGUEK-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results