Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. A25255ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25255ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25255ML Supplier TCI America Supplier No. A25255ML

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38793-01-0
Molecular Formula C11H23N
Molecular Weight (g/mol) 169.312
MDL Number MFCD07778407
InChI Key NQEVYZKSODGUEK-UHFFFAOYSA-N
Synonym 4-Pentylcyclohexylamine, 1-Amino-4-amylcyclohexane, 1-Amino-4-pentylcyclohexane
PubChem CID 13783029
IUPAC Name 4-pentylcyclohexan-1-amine
SMILES CCCCCC1CCC(CC1)N

Specifications

Color Colorless
Boiling Point 76°C
Quantity 5 mL
UN Number 2735
Formula Weight 169.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amylcyclohexylamine (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.