accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38821-53-3

IUPAC Name: (6R,7R)-7-[(2R)-2-amino-2-(cyclohexa-1,4-dien-1-yl)acetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid dihydrate
Synonyms: cefradine
Molecular Weight (g/mol): 385.44
Molecular Formula: C16H23N3O6S
InChi Key: HBKDHIDYMXXJLI-OEDJVVDHSA-N
SMILES: O.O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CCC=CC1)C2=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)