Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefradine, G-Biosciences
SCI_ED

Catalog No. S965101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S965101 1 g
S000006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S965101 Supplier G-Biosciences Supplier No. RC943
Add to Cart
Add to Cart

Cefradine (also known as cephradine) is a first generation cephalosporin antibiotic similar to cephalexin. It targets both gram-negative and gram-positive bacteria.

Cefradine has been used to study penicillin binding proteins, especially PBP3.

Chemical Identifiers

CAS 38821-53-3
Molecular Weight (g/mol) 349.40 g/mol

Specifications

Technique HPLC, UV/Vis, Spectrometry, Ion Chromatography, Gas Chromatography
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Multi-component Standard
Element Multi-element
Industry Type Biotech
Quantity 1 g
Percent Purity >97%
Grade Molecular Biology
Chemical Name or Material Cefradine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.