accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOO(S)Cl

CAS RN 3883-43-0

IUPAC Name: (2S)-2,3-dichloro-1,4-dioxane
Synonyms: (2S)-2,3-dichloro-1,4-dioxane
Molecular Weight (g/mol): 156.99
Molecular Formula: C4H6Cl2O2
InChi Key: ZOZUXFQYIYUIND-SYPWQXSBSA-N
SMILES: ClC1OCCO[C@H]1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)