Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2,3-Dichloro-1,4-dioxane 96.0+%, TCI America™
SDP

Catalog No. D22795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22795G 5 g
D227925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22795G Supplier TCI America Supplier No. D22795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3883-43-0
Molecular Formula C4H6Cl2O2
Molecular Weight (g/mol) 156.99
MDL Number MFCD00191421
InChI Key ZOZUXFQYIYUIND-SYPWQXSBSA-N
Synonym trans-2,3-Dichloro-1,4-diethylene Dioxide
PubChem CID 21766875
IUPAC Name (2S)-2,3-dichloro-1,4-dioxane
SMILES ClC1OCCO[C@H]1Cl

Specifications

Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Boiling Point 82°C
Formula Weight 156.99
Percent Purity ≥96.0% (GC)
Chemical Name or Material trans-2,3-Dichloro-1,4-dioxane
RTECSNumber JG9800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.