accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COO(S)NHOOOO

CAS RN 3886-08-6

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate
Molecular Weight (g/mol): 336.37
Molecular Formula: C17H22NO6
InChi Key: GLMODRZPPBZPPB-ZDUSSCGKSA-M
SMILES: CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)