Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-tert-Butyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™
SDP

Catalog No. C17155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17155G Supplier TCI America Supplier No. C17155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3886-08-6
Molecular Formula C17H22NO6
Molecular Weight (g/mol) 336.37
MDL Number MFCD00038274
InChI Key GLMODRZPPBZPPB-ZDUSSCGKSA-M
Synonym z-glu otbu-oh, cbz-glu obut-oh, cbz-l-glu otbu-oh, s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, n-cbz-l-glutamic acid 5-tert-butyl ester, n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester, 2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, 2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid, 5-tert-butyl n-carbobenzoxy-l-glutamate, pubchem6261
PubChem CID 107472
IUPAC Name (2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 87°C
Color White
Quantity 5 g
Formula Weight 337.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-tert-Butyl N-Carbobenzoxy-L-glutamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.