accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38862-25-8

H3CCH2OONOO

CAS RN 38862-25-8

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: ACGJEMXWUYWELU-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)